siRNA Details
Gene Symbol: Mesdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013149.1 | NP_001013167.1 | 1 | 1330 | ||
GenBank mRNA | BC091313.1 | - | 1 | 1330 | 7312243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
