siRNA Details
Gene Symbol: Prss23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007691.1 | NP_001007692.1 | 2 | 1197 | ||
GenBank mRNA | BC079179.1 | - | 2 | 1197 | 7130532 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029614, BC018517, AK156348, AK033671, AK009847, and others.
Gene Ontology Categories:
