siRNA Details
Gene Symbol: Usp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759807.1 | XP_008758029.1 | 11 | 2944 | ||
XM_238899.7 | XP_238899.4 | 10 | 2747 |

More Information
Other Species Targeted

Gene Ontology Categories:
