siRNA Details
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191629.1 | NP_001178558.1 | 23 | 2865 | ||
GenBank mRNA | FQ218032.1 | - | 21 | 2518 |

More Information
Other Species Targeted

Gene Ontology Categories:
