siRNA Details
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107536.1 | NP_001101006.1 | 4 | 497 | ||
XM_006229763.1 | XP_006229825.1 | 4 | 494 | |||
GenBank mRNA | BC169073.1 | - | 4 | 444 |
