siRNA Details
Gene Symbol: Chrdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107537.1 | NP_001101007.1 | 8 | 1084 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133709, BC019399, AK007577, AF338222.
Gene Ontology Categories:
