siRNA Details
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005885.2 | NP_001005885.1 | 5 | 522 | ||
XM_006229766.2 | XP_006229828.1 | 5 | 587 | |||
GenBank mRNA | AY325765.1 | - | 5 | 562 | ||
AY380804.1 | - | 5 | 580 | |||
BC098002.1 | - | 5 | 522 | 7453525 |
