siRNA Details
Gene Symbol: Numa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100691.1 | NP_001094161.1 | 13 | 4377 | ||
XM_008759723.1 | XP_008757945.1 | 13 | 4212 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Numa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100691.1 | NP_001094161.1 | 13 | 4377 | ||
XM_008759723.1 | XP_008757945.1 | 13 | 4212 |