siRNA Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107540.2 | NP_001101010.1 | 4, 5 | 406 | ||
XM_006229878.2 | XP_006229940.1 | 4, 5 | 631 | |||
XM_006229879.2 | XP_006229941.1 | 5, 6 | 749 |

More Information
Other Species Targeted

Gene Ontology Categories:
