siRNA Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107540.2 | NP_001101010.1 | 5 | 426 | ||
XM_006229878.2 | XP_006229940.1 | 5 | 651 | |||
XM_006229879.2 | XP_006229941.1 | 6 | 769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
