siRNA Details
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006997.1 | NP_001006998.1 | 5 | 1551 | ||
GenBank mRNA | BC083780.1 | - | 5 | 1551 | 7096039 |

More Information
Other Species Targeted

Gene Ontology Categories:
