siRNA Details
Gene Symbol: Egln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772679.1 | XP_008770901.1 | 3 | 1493 | ||
GenBank mRNA | AY228140.1 | - | 1 | 278 | ||
BC081694.1 | - | 1 | 522 |

More Information
Other Species Targeted

Gene Ontology Categories:
