siRNA Details
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100582.1 | NP_001094052.1 | 10 | 1170 | ||
XM_006229953.2 | XP_006230015.1 | 8 | 1080 | |||
XM_006229955.2 | XP_006230017.1 | 6 | 1474 | |||
XM_008759733.1 | XP_008757955.1 | 10 | 1142 | |||
XM_008759734.1 | XP_008757956.1 | 11 | 1500 | |||
XM_008759735.1 | XP_008757957.1 | 9 | 1263 | |||
XM_008759736.1 | XP_008757958.1 | 7 | 1500 | |||
GenBank mRNA | AY754026.1 | - | 5 | 388 |
