siRNA Details
Gene Symbol: Dennd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107546.2 | NP_001101016.2 | 14 | 2830 | ||
GenBank mRNA | BC168925.1 | - | 1 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
