siRNA Details
Gene Symbol: Pdzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024777.2 | NP_001019948.2 | 4 | 410 | ||
XM_008759741.1 | XP_008757963.1 | 3 | 2645 | |||
XM_008759742.1 | XP_008757964.1 | 4 | 615 | |||
XM_008759743.1 | XP_008757965.1 | 3 | 321 | |||
XM_008759744.1 | XP_008757966.1 | 5 | 2869 | |||
GenBank mRNA | AY387091.1 | - | 4 | 410 | ||
BC097426.1 | - | 3 | 307 | 7455141 |

More Information
Other Species Targeted

