siRNA Details
Gene Symbol: Tmem219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748988.3 | XP_003749036.1 | 6 | 1168 | ||
XM_008759928.1 | XP_008758150.1 | 5 | 854 | |||
XM_008759929.1 | XP_008758151.1 | 4 | 659 | |||
XM_008759930.1 | XP_008758152.1 | 4 | 656 | |||
GenBank mRNA | BC166576.1 | - | 5 | 722 | 7321704 | |
FQ216353.1 | - | 2 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
