siRNA Details
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203335.2 | NP_976080.1 | 2 | 272 | ||
GenBank mRNA | AY423047.1 | - | 2 | 255 | ||
BC166413.1 | - | 268 | 9100951 | |||
CB314647.1 | - | 2 | 288 | |||
FQ210919.1 | - | 2 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
