siRNA Details
Gene Symbol: Mcmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039608.2 | NP_001034697.2 | 14 | 1967 | ||
GenBank mRNA | BC111072.1 | - | 1 | 211 | 7445757 | |
BC160884.1 | - | 14 | 1967 | 7383399 | ||
FQ225204.1 | - | 5 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
