siRNA Details
Gene Symbol: Mcmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039608.2 | NP_001034697.2 | 14, 15 | 1999 | ||
GenBank mRNA | BC111072.1 | - | 1, 2 | 243 | 7445757 | |
BC160884.1 | - | 14, 15 | 1999 | 7383399 | ||
FQ225204.1 | - | 5, 6 | 705 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK196573.
Gene Ontology Categories:
