siRNA Details
Gene Symbol: RGD1307235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134546.1 | NP_001128018.1 | 1, 2 | 772 | ||
GenBank mRNA | FQ230810.1 | - | 1, 2 | 756 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029349, NM_173187, BC117950, AK129368, BC117949, and others.
