siRNA Details
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107560.2 | NP_001101030.1 | 11 | 1914 | ||
XM_006230457.1 | XP_006230519.1 | 11 | 1924 | |||
XM_006230458.1 | XP_006230520.1 | 12 | 2101 | |||
XM_006230459.2 | XP_006230521.1 | 13 | 2235 | |||
XM_006230460.1 | XP_006230522.1 | 12 | 2087 | |||
XM_006230461.1 | XP_006230523.1 | 8 | 1337 | |||
GenBank mRNA | BC166857.1 | - | 11 | 1914 | 9100036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
