siRNA Details
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107560.2 | NP_001101030.1 | 5 | 766 | ||
XM_006230457.1 | XP_006230519.1 | 5 | 776 | |||
XM_006230458.1 | XP_006230520.1 | 6 | 953 | |||
XM_006230459.2 | XP_006230521.1 | 7 | 1087 | |||
XM_006230460.1 | XP_006230522.1 | 6 | 939 | |||
XM_006230461.1 | XP_006230523.1 | 2 | 189 | |||
GenBank mRNA | BC166857.1 | - | 5 | 766 | 9100036 |
