siRNA Details
Gene Symbol: Scgb1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107561.1 | NP_001101031.1 | 2 | 271 | ||
GenBank mRNA | X60661.1 | - | 2 | 250 |

More Information
Other Species Targeted

