siRNA Details
Gene Symbol: Ifitm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760022.1 | XP_008758244.1 | 3 | 278 | ||
XM_008760023.1 | XP_008758245.1 | 2 | 230 | |||
XM_008760024.1 | XP_008758246.1 | 3 | 1560 | |||
XM_008760025.1 | XP_008758247.1 | 3 | 262 | |||
XM_219476.7 | XP_219476.5 | 2 | 1763 |
