siRNA Details
Gene Symbol: B4galnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107562.1 | NP_001101032.1 | 12, 13 | 1363 | ||
GenBank mRNA | FQ211564.1 | - | 12, 13 | 1195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
