siRNA Details
Gene Symbol: LOC100911365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749006.3 | XP_003749054.3 | 238 |
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107563.2 | NP_001101033.1 | 3 | 211 |
