siRNA Details
Gene Symbol: Tnpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100692.1 | NP_001094162.1 | 22 | 2548 | ||
XM_006231839.2 | XP_006231901.1 | 23 | 3139 | |||
XM_006231840.2 | XP_006231902.1 | 23 | 2863 | |||
XM_008760668.1 | XP_008758890.1 | 23 | 3151 | |||
GenBank mRNA | BC090323.1 | - | 20 | 2228 |
