siRNA Details
Gene Symbol: Mrgprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203470.1 | NP_982296.1 | 1 | 730 | ||
XM_008760116.1 | XP_008758338.1 | 2 | 954 | |||
XM_008760117.1 | XP_008758339.1 | 2 | 836 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
