siRNA Details
Gene Symbol: Mrpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107567.1 | NP_001101037.1 | 6 | 700 | ||
XM_008760120.1 | XP_008758342.1 | 6 | 602 |

More Information
Other Species Targeted

Gene Ontology Categories:
