siRNA Details
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006998.1 | NP_001006999.1 | 8 | 1132 | ||
XM_006230787.2 | XP_006230849.1 | 9 | 1435 | |||
XM_008760132.1 | XP_008758354.1 | 8 | 1622 | |||
XM_008760133.1 | XP_008758355.1 | 8 | 1622 | |||
XM_008760134.1 | XP_008758356.1 | 8 | 1116 | |||
GenBank mRNA | BC083850.1 | - | 8 | 1132 | 7122451 | |
FQ212890.1 | - | 2 | 247 |
