siRNA Details
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006998.1 | NP_001006999.1 | 6, 7 | 607 | ||
XM_006230787.2 | XP_006230849.1 | 7, 8 | 910 | |||
XM_008760132.1 | XP_008758354.1 | 6, 7 | 1097 | |||
XM_008760133.1 | XP_008758355.1 | 6, 7 | 1097 | |||
XM_008760134.1 | XP_008758356.1 | 6, 7 | 591 | |||
GenBank mRNA | BC083850.1 | - | 6, 7 | 607 | 7122451 |
