siRNA Details
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107569.1 | NP_001101039.1 | 5, 6 | 522 | ||
XM_008760137.1 | XP_008758359.1 | 5, 6 | 540 |

More Information
Other Species Targeted

Gene Ontology Categories:
