siRNA Details
Gene Symbol: Tmem151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107570.2 | NP_001101040.1 | 1 | 1458 | ||
XM_003749041.3 | XP_003749089.1 | 2 | 2109 | |||
GenBank mRNA | BC168914.1 | - | 1 | 1458 | 7313547 |
Gene Symbol: LOC103689952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230793.2 | XP_006230855.1 | 2090 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
