siRNA Details
Gene Symbol: LOC103689952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230793.2 | XP_006230855.1 | 483 |
Gene Symbol: Tmem151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749041.3 | XP_003749089.1 | 2 | 502 |

More Information
Other Species Targeted

