siRNA Details
Gene Symbol: Ovol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107572.1 | NP_001101042.1 | 4 | 917 | ||
XM_006230794.2 | XP_006230856.1 | 4 | 2941 | |||
XM_006230796.2 | XP_006230858.1 | 4 | 1601 | |||
XM_008760138.1 | XP_008758360.1 | 3 | 2057 |
