siRNA Details
Gene Symbol: Ehbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025117.2 | NP_001020288.2 | 16 | 2246 | ||
NM_001129997.1 | NP_001123469.1 | 16 | 5018 | |||
NM_001270704.1 | NP_001257633.1 | 16 | 2741 | |||
NM_001270705.1 | NP_001257634.1 | 15 | 2210 | |||
XM_008760139.1 | XP_008758361.1 | 16 | 2776 | |||
GenBank mRNA | BC087115.1 | - | 15 | 2173 | ||
BC129126.1 | - | 16 | 2246 | |||
BC167745.1 | - | 16 | 2522 | 7310903 |

More Information
Other Species Targeted

