siRNA Details
Gene Symbol: Mrpl49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047883.1 | NP_001041348.1 | 3 | 323 | ||
GenBank mRNA | AY325168.1 | - | 3 | 323 | ||
FQ209431.1 | - | 288 | ||||
FQ212546.1 | - | 302 | ||||
FQ235268.1 | - | 302 |
