siRNA Details
Gene Symbol: Mrpl49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047883.1 | NP_001041348.1 | 3 | 365 | ||
GenBank mRNA | AY325168.1 | - | 3 | 365 | ||
FQ209431.1 | - | 330 | ||||
FQ212546.1 | - | 344 | ||||
FQ235268.1 | - | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
