siRNA Details
Gene Symbol: Ppp2r5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181379.2 | NP_852044.1 | 2 | 269 | ||
XM_008760143.1 | XP_008758365.1 | 3 | 840 | |||
GenBank mRNA | AY251278.1 | - | 2 | 222 | ||
BC090324.1 | - | 2 | 269 | 7128996 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
