siRNA Details
Gene Symbol: Fermt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127543.1 | NP_001121015.1 | 7, 8 | 1008 | ||
XM_006230811.2 | XP_006230873.1 | 7, 8 | 1499 | |||
GenBank mRNA | BC166607.1 | - | 7, 8 | 1008 | 5598287 |
