siRNA Details
Gene Symbol: Atl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044241.1 | NP_001037706.1 | 4 | 720 | ||
XM_006230812.2 | XP_006230874.1 | 4 | 911 | |||
GenBank mRNA | DQ450893.1 | - | 4 | 720 | ||
FQ226220.1 | - | 4 | 537 | |||
FQ230464.1 | - | 4 | 788 | |||
FQ230893.1 | - | 4 | 788 |
