siRNA Details
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107575.1 | NP_001101045.1 | 7 | 633 | ||
XM_006230979.2 | XP_006231041.1 | 7 | 852 | |||
XM_006230981.2 | XP_006231043.1 | 7 | 795 | |||
XM_006230982.2 | XP_006231044.1 | 8 | 722 | |||
XM_006230983.2 | XP_006231045.1 | 7 | 633 | |||
XM_008760224.1 | XP_008758446.1 | 7 | 1104 | |||
GenBank mRNA | BC168911.1 | - | 7 | 748 | 7458553 |

More Information
Other Species Targeted

Gene Ontology Categories:
