siRNA Details
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107578.1 | NP_001101048.1 | 4 | 434 | ||
XM_006231089.2 | XP_006231151.1 | 5 | 606 | |||
GenBank mRNA | BC098745.1 | - | 5 | 497 | ||
FQ225563.1 | - | 5 | 557 | |||
FQ235337.1 | - | 4 | 371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
