siRNA Details
Gene Symbol: Glyatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126278.1 | NP_001119750.1 | 4 | 461 | ||
GenBank mRNA | BC160862.1 | - | 4 | 461 | 7323546 | |
FQ210014.1 | - | 615 | ||||
FQ219712.1 | - | 4 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
