siRNA Details
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100975.2 | NP_001094445.2 | 23 | 2464 | ||
XM_008760289.1 | XP_008758511.1 | 24 | 2763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
