siRNA Details
Gene Symbol: Pnma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107579.1 | NP_001101049.1 | 1 | 1243 | ||
XM_008773585.1 | XP_008771807.1 | 4 | 1581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
