siRNA Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013152.1 | NP_001013170.1 | 2 | 329 | ||
XM_006231208.2 | XP_006231270.1 | 6 | 775 | |||
GenBank mRNA | AY310155.1 | - | 765 | |||
AY325146.1 | - | 765 | ||||
BC091413.1 | - | 2 | 329 | 7326310 |
