siRNA Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013152.1 | NP_001013170.1 | 5 | 680 | ||
XM_006231208.2 | XP_006231270.1 | 9 | 1126 | |||
GenBank mRNA | BC091413.1 | - | 5 | 680 | 7326310 |

More Information
Other Species Targeted

Gene Ontology Categories:
