siRNA Details
Gene Symbol: Pdcd1lg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107582.2 | NP_001101052.2 | 4 | 820 | ||
XM_006231209.2 | XP_006231271.1 | 3 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
