siRNA Details
Gene Symbol: Gldc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107583.1 | NP_001101053.1 | 14 | 1977 | ||
XM_006231210.2 | XP_006231272.1 | 16 | 2123 | |||
XM_006231211.2 | XP_006231273.1 | 16 | 2124 |
